C12H316CE2EA Chemcia Scientific, LLC

6-Bromo-3H-thieno[3,2-d]pyrimidin-4-one

  • MDL number MFCD17677443
  • Cas Number : 215927-36-9
  • Molecular Weight : 231.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-bromothieno[3,2-d]pyrimidin-4-ol
Inchi Code 1S/C6H3BrN2OS/c7-4-1-3-5(11-4)6(10)9-2-8-3/h1-2H,(H,8,9,10)
Inchi Key IJSCGMBLRLDHPZ-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Tan Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=175

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=175
Availability details for
Quantity
  • Availability
  • Add to Cart