C12H316CE2F1 Chemcia Scientific, LLC

3-Benzyloxy-4-hydroxy-5-nitro-benzaldehyde

  • MDL number MFCD18909470
  • Cas Number : 312327-13-2
  • Molecular Weight : 273.24747

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=97

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=97
Availability details for
Quantity
  • Availability
  • Add to Cart