C12H316CE304 Chemcia Scientific, LLC

4,5,6,7-Hexahydro-benzo[d]isoxazole-3-carboxylic acid ethyl ester

  • MDL number MFCD08701222
  • Cas Number : 1013-14-5
  • Molecular Weight : 195.22001

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=167

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=167
Availability details for
Quantity
  • Availability
  • Add to Cart