C12H316CE306 Chemcia Scientific, LLC

Trifluoro-methanesulfonic acid 2-methoxy-4-formyl-6-nitro-phenyl ester

  • MDL number MFCD19441864
  • Cas Number : 1021493-52-6
  • Molecular Weight : 329.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-formyl-2-methoxy-6-nitrophenyl trifluoromethanesulfonate
Inchi Code 1S/C9H6F3NO7S/c1-19-7-3-5(4-14)2-6(13(15)16)8(7)20-21(17,18)9(10,11)12/h2-4H,1H3
Inchi Key UJHAJLHOIGFTGF-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=169

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=169
Availability details for
Quantity
  • Availability
  • Add to Cart