C12H316CE30C Chemcia Scientific, LLC

1,7-Dihydro-pyrrolo[2,3-d]pyrimidine-2,4-dione

  • MDL number MFCD08059284
  • Cas Number : 39929-79-8
  • Molecular Weight : 151.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1H-pyrrolo[2,3-d]pyrimidine-2,4(3H,5H)-dione
Inchi Code 1S/C6H5N3O2/c10-5-3-1-2-7-4(3)8-6(11)9-5/h1-2H,(H3,7,8,9,10,11)
Inchi Key HASUWNAFLUMMFI-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Tan Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=255

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=255
Availability details for
Quantity
  • Availability
  • Add to Cart