C12H316CE30E Chemcia Scientific, LLC

5-Dimethylamino-2-(2,2,2-trifluoro-acetyl)-penta-2,4-dienoic acid ethyl ester

  • MDL number MFCD19441866
  • Cas Number : 1146961-22-9
  • Molecular Weight : 265.23433

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Bright Orange Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=258

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=258
Availability details for
Quantity
  • Availability
  • Add to Cart