C12H316CE30F Chemcia Scientific, LLC

2-(2-Cyano-vinylamino)-malonic acid diethyl ester

  • MDL number MFCD19441867
  • Cas Number : 1021175-71-2
  • Molecular Weight : 226.23408

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Pink Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=261

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=261
Availability details for
Quantity
  • Availability
  • Add to Cart