C12H316CE310 Chemcia Scientific, LLC

5-Bromo-3-tert-butoxycarbonylamino-thiophene-2-carboxylic acid methyl ester

  • MDL number MFCD19441868
  • Cas Number : 946604-99-5
  • Molecular Weight : 336.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 5-bromo-3-[(tert-butoxycarbonyl)amino]-2-thiophenecarboxylate
Inchi Code 1S/C11H14BrNO4S/c1-11(2,3)17-10(15)13-6-5-7(12)18-8(6)9(14)16-4/h5H,1-4H3,(H,13,15)
Inchi Key OQCNMXAYFCAWPE-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Greenish Off-White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=264

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=264
Availability details for
Quantity
  • Availability
  • Add to Cart