C12H316CE311 Chemcia Scientific, LLC

2-Amino-5-methoxy-benzonitrile hydrochloride

  • MDL number MFCD18909469
  • Cas Number : 115661-38-6
  • Molecular Weight : 184.62673

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Salt Data HCl
Storage temp. 0~5
Physical Form Light Tan Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=105

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=105
Availability details for
Quantity
  • Availability
  • Add to Cart