C12H316CE313 Chemcia Scientific, LLC

Imidazo[1,2-a]pyridine-8-carboxylic acid methyl ester hydrochloride

  • MDL number MFCD19441984
  • Cas Number : 133427-07-3
  • Molecular Weight : 212.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl imidazo[1,2-a]pyridine-8-carboxylate hydrochloride
Inchi Code 1S/C9H8N2O2.ClH/c1-13-9(12)7-3-2-5-11-6-4-10-8(7)11;/h2-6H,1H3;1H
Inchi Key SPZWZNLBQSHSLQ-UHFFFAOYSA-N

Description

Purity 97%
Salt Data HCl
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=270

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=270
Availability details for
Quantity
  • Availability
  • Add to Cart