C12H316CE31A Chemcia Scientific, LLC

5-(2-Bromo-acetyl)-thiophene-2-carboxylic acid methyl ester

  • MDL number MFCD13191387
  • Cas Number : 4192-32-9
  • Molecular Weight : 263.11119

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=288

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=288
Availability details for
Quantity
  • Availability
  • Add to Cart