C12H316CE31B Chemcia Scientific, LLC

5-(2,2-Dibromo-acetyl)-thiophene-2-carboxylic acid methyl ester

  • MDL number MFCD19441877
  • Cas Number : 656227-47-3
  • Molecular Weight : 342.00722

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=362

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=362
Availability details for
Quantity
  • Availability
  • Add to Cart