C12H316CE31E Chemcia Scientific, LLC

2-Chloro-5-vinyl-pyridine

  • MDL number MFCD11870540
  • Cas Number : 157670-28-5
  • Molecular Weight : 139.58557

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. 0~5
Physical Form Yellow Viscous Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=293

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=293
Availability details for
Quantity
  • Availability
  • Add to Cart