C12H316CE320 Chemcia Scientific, LLC

8-Bromo-6-nitroimidazo[1,2-a]pyridine hydrochloride

  • MDL number MFCD09800973
  • Cas Number : 957120-43-3
  • Molecular Weight : 278.4938

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Salt Data HCl
Storage temp. 0~5
Physical Form Grey Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=299

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=299
Availability details for
Quantity
  • Availability
  • Add to Cart