C12H316CE321 Chemcia Scientific, LLC

2-(4-tert-Butyl-phenyl)-2-methyl-propionic acid methyl ester

  • MDL number MFCD19441870
  • Cas Number : 157444-69-4
  • Molecular Weight : 234.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-(4-tert-butylphenyl)-2-methylpropanoate
Inchi Code 1S/C15H22O2/c1-14(2,3)11-7-9-12(10-8-11)15(4,5)13(16)17-6/h7-10H,1-6H3
Inchi Key MFQRFTBOTSBMIO-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Off-White Crystalline Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=304

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=304
Availability details for
Quantity
  • Availability
  • Add to Cart