C12H316CE323 Chemcia Scientific, LLC

2-(4-Dimethylamino-phenyl)-2-methyl-propionic acid

  • MDL number MFCD10035606
  • Cas Number : 228411-16-3
  • Molecular Weight : 207.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-[4-(dimethylamino)phenyl]-2-methylpropanoic acid
Inchi Code 1S/C12H17NO2/c1-12(2,11(14)15)9-5-7-10(8-6-9)13(3)4/h5-8H,1-4H3,(H,14,15)
Inchi Key AJVDJWZAIIWTRR-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Light Grey Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=314

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=314
Availability details for
Quantity
  • Availability
  • Add to Cart