C12H316CE325 Chemcia Scientific, LLC

2-(4-tert-Butyl-phenyl)-2-methyl-propionic acid

  • MDL number MFCD09260840
  • Cas Number : 93748-14-2
  • Molecular Weight : 220.3143

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0-5*C
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=323

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=323
Availability details for
Quantity
  • Availability
  • Add to Cart