C12H316CE328 Chemcia Scientific, LLC

N-(2-Cyano-cyclohex-1-enyl)-acetamide

  • MDL number MFCD00102302
  • Cas Number : 500896-58-2
  • Molecular Weight : 164.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(2-cyanocyclohex-1-en-1-yl)acetamide
Inchi Code 1S/C9H12N2O/c1-7(12)11-9-5-3-2-4-8(9)6-10/h2-5H2,1H3,(H,11,12)
Inchi Key NJAAKBJNTCGTOC-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=331

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=331
Availability details for
Quantity
  • Availability
  • Add to Cart