C12H316CE331 Chemcia Scientific, LLC

N,N-Bis-boc-2-amino-6-fluorobenzonitrile

  • MDL number MFCD19441876
  • Cas Number : 1313712-57-0
  • Molecular Weight : 336.36632

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=359

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=359
Availability details for
Quantity
  • Availability
  • Add to Cart