C12H316CE334 Chemcia Scientific, LLC

6-Chloro-benzo[d]isoxazol-3-ylamine

  • MDL number MFCD09055056
  • Cas Number : 89692-53-5
  • Molecular Weight : 168.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-chloro-1,2-benzisoxazol-3-amine
Inchi Code 1S/C7H5ClN2O/c8-4-1-2-5-6(3-4)11-10-7(5)9/h1-3H,(H2,9,10)
Inchi Key VUMFEESWELAKKV-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=356

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=356
Availability details for
Quantity
  • Availability
  • Add to Cart