C12H316CE335 Chemcia Scientific, LLC

4-Methoxy-benzo[d]isoxazol-3-ylamine

  • MDL number MFCD03407354
  • Cas Number : 177995-40-3
  • Molecular Weight : 164.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-methoxy-1,2-benzisoxazol-3-ylamine
Inchi Code 1S/C8H8N2O2/c1-11-5-3-2-4-6-7(5)8(9)10-12-6/h2-4H,1H3,(H2,9,10)
Inchi Key YFNIGSJHKGKXEQ-UHFFFAOYSA-N

Description

Purity 90%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=358

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=358
Availability details for
Quantity
  • Availability
  • Add to Cart