C12H316CE33A Chemcia Scientific, LLC

2-Pyridin-2-yl-malonic acid di-tert-butyl ester

  • MDL number MFCD19441885
  • Cas Number : 1104643-39-1
  • Molecular Weight : 293.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name di(tert-butyl) 2-(2-pyridinyl)malonate
Inchi Code 1S/C16H23NO4/c1-15(2,3)20-13(18)12(11-9-7-8-10-17-11)14(19)21-16(4,5)6/h7-10,12H,1-6H3
Inchi Key BJPGCHIJQUEXDY-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Yellow Liquid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=379

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=379
Availability details for
Quantity
  • Availability
  • Add to Cart