C12H316CE33C Chemcia Scientific, LLC

2-Fluoro-4-hydrazino-benzonitrile

  • MDL number MFCD08689472
  • Cas Number : 129946-64-1
  • Molecular Weight : 151.14437

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=384

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=384
Availability details for
Quantity
  • Availability
  • Add to Cart