C12H316CE33D Chemcia Scientific, LLC

(2-Cyano-3-fluoro-phenyl)-carbamic acid tert-butyl ester

  • MDL number MFCD12078320
  • Cas Number : 1153762-99-2
  • Molecular Weight : 236.24801

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=368

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=368
Availability details for
Quantity
  • Availability
  • Add to Cart