C12H316CE33E Chemcia Scientific, LLC

(3-Amino-benzo[d]isoxazol-4-yl)-carbamic acid tert-butyl ester

  • MDL number MFCD19441887
  • Cas Number : 1313712-34-3
  • Molecular Weight : 249.27165

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=386

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=386
Availability details for
Quantity
  • Availability
  • Add to Cart