C12H316CE34C Chemcia Scientific, LLC

N-[6-(2-Bromo-acetyl)-pyridazin-3-yl]-acetamide

  • MDL number MFCD18910939
  • Cas Number : 1313712-11-6
  • Molecular Weight : 258.07586

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=110

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=110
Availability details for
Quantity
  • Availability
  • Add to Cart