C12H316CE34D Chemcia Scientific, LLC

6,7-Dihydro-5H-quinolin-8-one oxime

  • MDL number MFCD20921531
  • Cas Number : 58509-59-4
  • Molecular Weight : 162.19285

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. 0~5
Physical Form Dark Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=808

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=808
Availability details for
Quantity
  • Availability
  • Add to Cart