C12H316CE34F Chemcia Scientific, LLC

2,4,6,7-Tetrahydro-pyrazolo[4,3-c]pyridine-3,5-dicarboxylic acid 5-tert-butyl ester 3-ethyl ester

  • MDL number MFCD20275307
  • Cas Number : 518990-23-3
  • Molecular Weight : 295.34117

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Pale Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2275

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2275
Availability details for
Quantity
  • Availability
  • Add to Cart