C12H316CE356 Chemcia Scientific, LLC

5-[(5-Bromo-pyridin-2-ylamino)-methylene]-2,2-dimethyl-[1,3]dioxane-4, 6-dione

  • MDL number MFCD11999829
  • Cas Number : 25165-70-2
  • Molecular Weight : 327.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-{[(5-bromo-2-pyridinyl)amino]methylene}-2,2-dimethyl-1,3-dioxane-4,6-dione
Inchi Code 1S/C12H11BrN2O4/c1-12(2)18-10(16)8(11(17)19-12)6-15-9-4-3-7(13)5-14-9/h3-6H,1-2H3,(H,14,15)
Inchi Key QHFOXJGGDKYKFE-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Light Orange Crystalline Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=424

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=424
Availability details for
Quantity
  • Availability
  • Add to Cart