C12H316CE357 Chemcia Scientific, LLC

2-[(2,2-Dimethyl-4,6-dioxo-[1,3]dioxan-5-ylidenemethyl)-amino]-benzoic acid methyl ester

  • MDL number MFCD00472585
  • Cas Number : 185130-13-6
  • Molecular Weight : 305.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-{[(2,2-dimethyl-4,6-dioxo-1,3-dioxan-5-ylidene)methyl]amino}benzoate
Inchi Code 1S/C15H15NO6/c1-15(2)21-13(18)10(14(19)22-15)8-16-11-7-5-4-6-9(11)12(17)20-3/h4-8,16H,1-3H3
Inchi Key ZWEQGBYLZYNDNT-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=426

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=426
Availability details for
Quantity
  • Availability
  • Add to Cart