C12H316CE359 Chemcia Scientific, LLC

4-Hydroxy-quinoline-8-carboxylic acid methyl ester

  • MDL number MFCD18417588
  • Cas Number : 860206-84-4
  • Molecular Weight : 203.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 4-oxo-1,4-dihydro-8-quinolinecarboxylate
Inchi Code 1S/C11H9NO3/c1-15-11(14)8-4-2-3-7-9(13)5-6-12-10(7)8/h2-6H,1H3,(H,12,13)
Inchi Key VNLGUOZXFZJECD-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1192

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1192
Availability details for
Quantity
  • Availability
  • Add to Cart