C12H316CE35E Chemcia Scientific, LLC

4-Bromo-3-bis(tert-butoxycarbonyl)amino-thiophene-2-carboxylic acid methyl ester

  • MDL number MFCD19441891
  • Cas Number : 1313712-43-4
  • Molecular Weight : 436.32484

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. 0~5
Physical Form Light Tan Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=439

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=439
Availability details for
Quantity
  • Availability
  • Add to Cart