C12H316CE364 Chemcia Scientific, LLC

5-Amino-6-iodo-pyridine-2-carbonitrile

  • MDL number MFCD18260519
  • Cas Number : 1150542-25-8
  • Molecular Weight : 245.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-amino-6-iodo-2-pyridinecarbonitrile
Inchi Code 1S/C6H4IN3/c7-6-5(9)2-1-4(3-8)10-6/h1-2H,9H2
Inchi Key CSGBAAUMIULLPT-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Light Brown Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=444

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=444
Availability details for
Quantity
  • Availability
  • Add to Cart