C12H316CE369 Chemcia Scientific, LLC

5-[(2-Bromo-phenylamino)-methylene]-2,2-dimethyl-[1,3]dioxane-4,6-dione

  • MDL number MFCD08582139
  • Cas Number : 187278-04-2
  • Molecular Weight : 326.14889

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=448

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=448
Availability details for
Quantity
  • Availability
  • Add to Cart