C12H316CE36D Chemcia Scientific, LLC

4-[(2,2-Dimethyl-4,6-dioxo-[1,3]dioxan-5-ylidenemethyl)-amino]-benzoic acid ethyl ester

  • MDL number MFCD02936594
  • Cas Number : 205810-04-4
  • Molecular Weight : 319.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 4-{[(2,2-dimethyl-4,6-dioxo-1,3-dioxan-5-ylidene)methyl]amino}benzoate
Inchi Code 1S/C16H17NO6/c1-4-21-13(18)10-5-7-11(8-6-10)17-9-12-14(19)22-16(2,3)23-15(12)20/h5-9,17H,4H2,1-3H3
Inchi Key ZMPTVSZNLHVTRO-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Yellow Crystalline Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=450

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=450
Availability details for
Quantity
  • Availability
  • Add to Cart