C12H316CE372 Chemcia Scientific, LLC

4-Hydroxy-quinoline-6-carboxylic acid

  • MDL number MFCD16987424
  • Cas Number : 1065092-81-0
  • Molecular Weight : 189.17219

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=370

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=370
Availability details for
Quantity
  • Availability
  • Add to Cart