C12H316CE376 Chemcia Scientific, LLC

2-Amino-6,7-dihydro-5H-thiazolo[5,4-b]pyridine-4-carboxylic acid tert-butyl ester

  • MDL number MFCD19441894
  • Cas Number : 1313712-23-0
  • Molecular Weight : 255.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 2-amino-6,7-dihydro[1,3]thiazolo[5,4-b]pyridine-4(5H)-carboxylate
Inchi Code 1S/C11H17N3O2S/c1-11(2,3)16-10(15)14-6-4-5-7-8(14)17-9(12)13-7/h4-6H2,1-3H3,(H2,12,13)
Inchi Key LZNIMGWYMCZLLL-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=452

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=452
Availability details for
Quantity
  • Availability
  • Add to Cart