C12H316CE379 Chemcia Scientific, LLC

5,6-Dihydro-4H-cyclopentathiazol-2-ylamine hydrobromide

  • MDL number MFCD19441980
  • Cas Number : 927682-21-1
  • Molecular Weight : 221.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5,6-dihydro-4H-cyclopenta[d][1,3]thiazol-2-amine hydrobromide
Inchi Code 1S/C6H8N2S.BrH/c7-6-8-4-2-1-3-5(4)9-6;/h1-3H2,(H2,7,8);1H
Inchi Key WMNZJWNOIHWFSQ-UHFFFAOYSA-N

Description

Purity 97%
Salt Data HBr
Storage temp. 0~5
Physical Form Purple-Like Tan Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=475

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=475
Availability details for
Quantity
  • Availability
  • Add to Cart