C12H316CE384 Chemcia Scientific, LLC

[1,2,4]Triazolo[1,5-a]pyridin-2-ylamine

  • MDL number MFCD11846603
  • Cas Number : 874-46-4
  • Molecular Weight : 134.14152

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=486

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=486
Availability details for
Quantity
  • Availability
  • Add to Cart