C12H316CE38E Chemcia Scientific, LLC

2-(6-Fluoro-pyridin-2-ylamino)-ethanol

  • MDL number MFCD14600159
  • Cas Number : 1000981-41-8
  • Molecular Weight : 156.16098

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Pale Yellow Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=497

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=497
Availability details for
Quantity
  • Availability
  • Add to Cart