C12H316CE390 Chemcia Scientific, LLC

1,2,3,4-Tetrahydro-pyrido[1,2-a]pyrimidin-6-one hydrochloride

  • MDL number MFCD23115562
  • Cas Number : 1332586-91-0
  • Molecular Weight : 186.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3,4-dihydro-1H-pyrido[1,2-a]pyrimidin-6(2H)-one hydrochloride
Inchi Code 1S/C8H10N2O.ClH/c11-8-4-1-3-7-9-5-2-6-10(7)8;/h1,3-4,9H,2,5-6H2;1H
Inchi Key BCBBDHFUWJLGSF-UHFFFAOYSA-N

Description

Purity 95%
Salt Data HCl
Storage temp. 0~5
Physical Form Purple Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=499

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=499
Availability details for
Quantity
  • Availability
  • Add to Cart