C12H316CE396 Chemcia Scientific, LLC

2,3-Dihydro-1H-imidazo[1,2-a]pyridin-5-one hydrochloride

  • MDL number MFCD19441988
  • Cas Number : 1332581-57-3
  • Molecular Weight : 172.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,3-dihydroimidazo[1,2-a]pyridin-5(1H)-one hydrochloride
Inchi Code 1S/C7H8N2O.ClH/c10-7-3-1-2-6-8-4-5-9(6)7;/h1-3,8H,4-5H2;1H
Inchi Key RZGKBSSATKJQCB-UHFFFAOYSA-N

Description

Purity 95%
Salt Data HCl
Storage temp. 0~5
Physical Form Grey Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=505

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=505
Availability details for
Quantity
  • Availability
  • Add to Cart