C12H316CE397 Chemcia Scientific, LLC

(S)-2-(2-Fluoro-benzyl)-3-hydroxy-5-oxo-2,5-dihydro-pyrrole-1-carboxylic acid tert-butyl ester

  • MDL number MFCD19441910
  • Cas Number : 1332581-53-9
  • Molecular Weight : 307.32456

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=509

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=509
Availability details for
Quantity
  • Availability
  • Add to Cart