C12H316CE39E Chemcia Scientific, LLC

6-Bromo-imidazo[1,2-a]pyridine-2-carbaldehyde

  • MDL number MFCD06739238
  • Cas Number : 885276-09-5
  • Molecular Weight : 225.04585

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Brown Crystalline Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=518

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=518
Availability details for
Quantity
  • Availability
  • Add to Cart