C12H316CE3AA Chemcia Scientific, LLC

5-Methyl-imidazo[1,2-a]pyridine-2-carbaldehyde

  • MDL number MFCD05863387
  • Cas Number : 118000-44-5
  • Molecular Weight : 160.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-methylimidazo[1,2-a]pyridine-2-carbaldehyde
Inchi Code 1S/C9H8N2O/c1-7-3-2-4-9-10-8(6-12)5-11(7)9/h2-6H,1H3
Inchi Key WNIYPDOZDBSIIE-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=535

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=535
Availability details for
Quantity
  • Availability
  • Add to Cart