C12H316CE3AC Chemcia Scientific, LLC

3-Bromo-8-methyl-imidazo[1,2-a]pyridine-2-carbaldehyde

  • MDL number MFCD19441928
  • Cas Number : 175878-06-5
  • Molecular Weight : 239.07294

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=537

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=537
Availability details for
Quantity
  • Availability
  • Add to Cart