C12H316CE3B1 Chemcia Scientific, LLC

6-Bromo-imidazo[1,2-a]pyridine-2-carboxylic acid ethyl ester hydrobromide

  • MDL number MFCD09991793
  • Cas Number : 1177092-98-6
  • Molecular Weight : 350.0114

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Salt Data HBr
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=542

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=542
Availability details for
Quantity
  • Availability
  • Add to Cart