C12H316CE3B5 Chemcia Scientific, LLC

3-Bromo-7-methyl-imidazo[1,2-a]pyridine-2-carboxylic acid ethyl ester hydrobromide

  • MDL number MFCD19442000
  • Cas Number : 1332588-44-9
  • Molecular Weight : 364.03849

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Salt Data HBr
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=546

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=546
Availability details for
Quantity
  • Availability
  • Add to Cart