C12H316CE3B6 Chemcia Scientific, LLC

3-Bromo-8-methyl-imidazo[1,2-a]pyridine-2-carboxylic acid ethyl ester hydrobromide

  • MDL number MFCD19442001
  • Cas Number : 1332589-38-4
  • Molecular Weight : 364.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 3-bromo-8-methylimidazo[1,2-a]pyridine-2-carboxylate hydrobromide
Inchi Code 1S/C11H11BrN2O2.BrH/c1-3-16-11(15)8-9(12)14-6-4-5-7(2)10(14)13-8;/h4-6H,3H2,1-2H3;1H
Inchi Key NMQGMBWLCOWKOD-UHFFFAOYSA-N

Description

Purity 97%
Salt Data HBr
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=547

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=547
Availability details for
Quantity
  • Availability
  • Add to Cart