C12H316CE3C0 Chemcia Scientific, LLC

2-Bromo-[1,2,4]triazolo[1,5-a]pyridine

  • MDL number MFCD19441933
  • Cas Number : 1021019-03-3
  • Molecular Weight : 198.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0-5C
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=558

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=558
Availability details for
Quantity
  • Availability
  • Add to Cart